Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555452 Supplier Invitrogen™ Supplier No. PA555452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAGEVVACLI RVPSEVVKQR AQVSASTRTF QIFSNILYEE GIQGLYRGYK STVLREIPFS LVQFPLWESL KALWSWRQDH VVDSWQSAVC GAFAGGFAAA VTTPLDVAKT RITLAKAGSS TADGNVLSVL HGV Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Mitochondrial carriers, including SLC25A26, are a family of transport proteins found mostly in the inner membranes of mitochondria. They shuttle metabolites and cofactors through the mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A26
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A26
Gene Accession No. Q70HW3
Gene Alias 4930433D19Rik; 4933433F13Rik; AW557176; COXPD28; D6Bwg0781e; hypothetical protein LOC560478; mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; samc; SLC25A26; Slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; solute carrier family 25 member 26; solute carrier family 25, member 26; SZFULLNAME2; SZGENESYMBOL2; zgc:110080
Gene Symbols SLC25A26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A26. Recombinant protein control fragment (Product #RP-90315).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115286
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.