Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120376 Supplier Invitrogen™ Supplier No. PA5120376
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis, Mouse brain. The target is usually found in the following locations: Mitochondrion inner membrane, Multi-pass membrane protein.

Mitochondrial carriers, including SLC25A26, are a family of transport proteins found mostly in the inner membranes of mitochondria. They shuttle metabolites and cofactors through the mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A26
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC25A26
Gene Accession No. Q5U680
Gene Alias 4930433D19Rik; 4933433F13Rik; AW557176; COXPD28; D6Bwg0781e; hypothetical protein LOC560478; mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; samc; SLC25A26; Slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; solute carrier family 25 member 26; solute carrier family 25, member 26; SZFULLNAME2; SZGENESYMBOL2; zgc:110080
Gene Symbols SLC25A26
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67582
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.