Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A26 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4376 Supplier Bioss Supplier No. BS19813R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mitochondrial carriers, including SLC25A26, are a family of transport proteins found mostly in the inner membranes of mitochondria. They shuttle metabolites and cofactors through the mitochondrial membrane.

Specifications

Antigen SLC25A26
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC25A26
Gene Accession No. Q70HW3
Gene Alias 4930433D19Rik; 4933433F13Rik; AW557176; BOS_20856; COXPD28; D6Bwg0781e; hypothetical protein LOC560478; LOC100338902; Mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; samc; SLC25A26; Slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; Solute carrier family 25 member 26; solute carrier family 25, member 26; SZFULLNAME2; SZGENESYMBOL2; zgc:110080
Gene Symbols SLC25A26
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC25A26.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115286
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.