Learn More
Invitrogen™ SLC25A28 Polyclonal Antibody

Description
Peptide sequence: NTQESLALNS HITGHITGMA SAFRTVYQVG GVTAYFRGVQ ARVIYQIPST Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
Specifications
Specifications
| Antigen | SLC25A28 |
| Applications | Immunohistochemistry, Western Blot, Surface Plasmon Resonance |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SLC25A28 |
| Gene Accession No. | Q96A46 |
| Gene Alias | 2210403D18Rik; DKFZp547C109; hMRS3/4; Mfrn2; Mitochondrial iron transporter 2; mitochondrial RNA splicing protein 3/4; mitochondrial RNA-splicing protein 3/4 homolog; mitoferrin 2; mitoferrin-2; Mitoferrin-2-like protein; MRS3/4; MRS4L; NPD016; putative mitochondrial solute carrier; SLC25A28; solute carrier family 25 (mitochondrial iron transporter), member 28; solute carrier family 25 member 28; solute carrier family 25, member 28 |
| Gene Symbols | SLC25A28 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.