Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A28 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA531839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531839 Supplier Thermo Scientific Supplier No. PA531839

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1. Predicted reactivity: Mouse (98%), Rat (97%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Mitochondrial iron transporter that mediates iron uptake. Probably required for heme synthesis of hemoproteins and Fe-S cluster assembly in non-erythroid cells. The iron delivered into the mitochondria, presumably as Fe(2+), is then probably delivered to ferrochelatase to catalyze Fe(2+) incorporation into protoprophyrin IX to make heme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A28
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SLC25A28
Gene Accession No. Q96A46
Gene Alias 2210403D18Rik; DKFZp547C109; hMRS3/4; Mfrn2; Mitochondrial iron transporter 2; mitochondrial RNA splicing protein 3/4; mitochondrial RNA-splicing protein 3/4 homolog; mitoferrin 2; mitoferrin-2; Mitoferrin-2-like protein; MRS3/4; MRS4L; NPD016; putative mitochondrial solute carrier; SLC25A28; solute carrier family 25 (mitochondrial iron transporter), member 28; solute carrier family 25 member 28; solute carrier family 25, member 28
Gene Symbols SLC25A28
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 142 and 364 of Human SLC25A28
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.