Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556556 Supplier Invitrogen™ Supplier No. PA556556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDSPLNQFLA GAAAGAIQCV ICCPMELAKT RLQLQDAGPA RTYKGSLDCL AQIYGHEGLR GVNRGMVSTL LRETPSFGVY FLTYDALTRA LGCEPGDRLL VPKLLLAGGT SGIVSWLSTY PVDVV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

SLC25A29 belongs to the mitochondrial carrier family and it may has palmitoylcarnitine transporting activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A29
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc25a29
Gene Accession No. Q8N8R3
Gene Alias C030003J19Rik; C14orf69; CACL; CACT-like; carnitine/acylcarnitine translocase-like; carnitine-acylcarnitine translocase like; hypothetical protein LOC569608; LOC100715169; LOW QUALITY PROTEIN: mitochondrial basic amino acids transporter; mCACL; mitchondrial carnitine/acylcarnitine carrier protein CACL; mitochondrial basic amino acids transporter; Mitochondrial carnitine/acylcarnitine carrier protein CACL; mitochondrial ornithine transporter 3; Ornt3; Slc25a29; solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29; solute carrier family 25 (mitochondrial carrier, palmitoylcarnitine transporter), member 29; solute carrier family 25 member 29; solute carrier family 25, member 29; zgc:114094
Gene Symbols Slc25a29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A29. Recombinant protein control fragment (Product #RP-89069).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123096
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.