Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544133 Supplier Invitrogen™ Supplier No. PA544133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LAGGVAAAVS KTAVAPIERV KLLLQVQASS KQISPEARYK GMVDCLVRIP Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Mitochondrial ADP/ATP carriers, such as SLC25A31, are nuclear-coded mitochondrial proteins that catalyze the exchange of ATP generated in mitochondria by ATP synthase (see MIM 108729) against ADP produced in cytosol by most energy-consuming reactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A31
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC25A31
Gene Accession No. Q9H0C2
Gene Alias 1700034J06Rik; Aac4; adenine nucleotide translocase 4; adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 4; ADP/ATP translocase 4; ANT 4; Ant4; LOC541168 protein; Sfec; SFEC35kDa; SLC25A31; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 member 31; Sperm flagellar energy carrier protein
Gene Symbols SLC25A31
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC25A31 (aa 25-74).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83447
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.