Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582520
Description
Immunogen sequence: FARTRLGVDI GKGPEERQFK GLGDCIMKIA KSDGIAGLYQ GFGVSVQGII VYRA.
Mitochondrial ADP/ATP carriers, such as SLC25A31, are nuclear-coded mitochondrial proteins that catalyze the exchange of ATP generated in mitochondria by ATP synthase (see MIM 108729) against ADP produced in cytosol by most energy-consuming reactions.
Specifications
SLC25A31 | |
Polyclonal | |
Unconjugated | |
SLC25A31 | |
1700034J06Rik; Aac4; adenine nucleotide translocase 4; adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 4; ADP/ATP translocase 4; ANT 4; Ant4; LOC541168 protein; Sfec; SFEC35kDa; SLC25A31; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 member 31; Sperm flagellar energy carrier protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83447 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0C2 | |
SLC25A31 | |
Recombinant protein corresponding to Human SLC25A31. Recombinant protein control fragment (Product #RP-90992). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction