Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542496 Supplier Invitrogen™ Supplier No. PA542496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VRYENLIAGV SGGVLSNLAL HPLDLVKIRF AVSDGLELRP KYNGILHCLT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

SLC25A32 transports folate across the inner membranes of mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A32
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC25A32
Gene Accession No. Q9H2D1
Gene Alias 2610043O12Rik; MFT; MFTC; mitochondrial folate transporter/carrier; RGD1565789; RREI; SLC25A32; solute carrier family 25 (mitochondrial folate carrier) , member 32; solute carrier family 25 (mitochondrial folate carrier), member 32; solute carrier family 25 member 32; solute carrier family 25, member 32
Gene Symbols SLC25A32
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC25A32 (aa 21-70).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81034
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.