Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521793 Supplier Invitrogen™ Supplier No. PA521793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (87%), Rat (88%), Sheep (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The SLC25A33 gene belongs to the solute carrier family 25 (SLC25), which is the largest family of mitochondrial transport proteins comprising 53 members. The SLC25A33 gene encodes a mitochondrial transporter responsible for the transport of pyrimidine nucleotides, specifically deoxyadenine nucleotides, across the inner mitochondrial membrane. This transport is crucial for nucleotide salvage pathways, ensuring the synthesis and maintenance of mitochondrial DNA. SLC25A33 also plays a significant role in cellular energy metabolism, linking cytosolic and mitochondrial nucleotide pools. Mutations or dysregulation of this gene can impact mitochondrial function and have been implicated in various metabolic disorders and mitochondrial diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A33
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SLC25A33
Gene Accession No. Q9BSK2
Gene Alias BMSC-MCP; Bone marrow stromal cell mitochondrial carrier protein; huBMSC-MCP; mitochondrial carrier protein; novel mitochondrial carrier protein; PNC1; PNC1 protein; Protein PNC1; SLC25A33; solute carrier family 25 (pyrimidine nucleotide carrier), member 33; solute carrier family 25 member 33; solute carrier family 25, member 33
Gene Symbols SLC25A33
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC25A33. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84275
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.