Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562466 Supplier Invitrogen™ Supplier No. PA562466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IYMVKTHLQA QAASEIAVGH QYKHQGMFQA LTEIGQKHGL VGLWRGALGG LPRVIVGSST QLCTFSSTKD LLSQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

SLC25A34 belongs to the SLC25 family of mitochondrial carrier proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A35
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A35
Gene Accession No. Q3KQZ1
Gene Alias 1810012H11Rik; si:ch211-268m12.5; similar to 1810012H11Rik; slc25a35; Solute carrier family 25 member 35; solute carrier family 25, member 35
Gene Symbols SLC25A35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A35. Recombinant protein control fragment (Product #RP-93246).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.