Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526720
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526720 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526720 Supplier Invitrogen™ Supplier No. PA526720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A37 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A37
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SLC25A37
Gene Accession No. Q66H23, Q9NYZ2
Gene Alias 1700020E22Rik; 4930513O14Rik; 4930526G11Rik; AI848481; C330015G08Rik; frascati; HT015; Mfrn; Mfrn1; mitochondrial carrier domain containing protein RGD1359361; mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitochondrial solute carrier-like protein; mitoferrin; mitoferrin 1; mitoferrin-1; MSC; MSCP; predicted protein of HQ2217; PRO1278; PRO1584; PRO2217; RGD1359361; Slc25a37; solute carrier family 25 (mitochondrial iron transporter), member 37; solute carrier family 25 member 37; solute carrier family 25, member 37
Gene Symbols SLC25A37
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 309-338 amino acids from the C-terminal region of human SLC25A37.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306000, 51312
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.