Learn More
Invitrogen™ SLC25A38 Polyclonal Antibody

Description
Positive Samples: 293T; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein Immunogen sequence: MIQNSRPSLL QPQDVGDTVE TLMLHPVIKA FLCGSISGTC STLLFQPLDL LKTRLQTLQP SDHGSRRVGM LAVLLKVVRT ESLLGLWKGM SPSIVRCVPG.
Specifications
Specifications
| Antigen | SLC25A38 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.72 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | SLC25A38 |
| Gene Accession No. | Q91XD8, Q96DW6 |
| Gene Alias | appoptosin; AV019094; BC010801; HAMAP-Rule:MF_03064}; hypothetical protein FLJ20551; mitochondrial glycine transporter; Mitochondrial glycine transporter GlyC; RGD1311914; SIDBA2; Slc25a38; solute carrier family 25 member 38; solute carrier family 25 member 38 {ECO:0000255; solute carrier family 25, member 38 |
| Gene Symbols | SLC25A38 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.