Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542472
Description
Peptide sequence: VGIYFGTLYS LKQYFLRGHP PTALESVMLG VGSRSVAGVC MSPITVIKTR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.
SLC25A38 is a members of the solute carrier family 25 (SLC25) which is known to transport molecules over the mitochondrial membrane.
Specifications
SLC25A38 | |
Polyclonal | |
Unconjugated | |
SLC25A38 | |
appoptosin; AV019094; BC010801; HAMAP-Rule:MF_03064}; hypothetical protein FLJ20551; mitochondrial glycine transporter; Mitochondrial glycine transporter GlyC; RGD1311914; SIDBA2; Slc25a38; solute carrier family 25 member 38; solute carrier family 25 member 38 {ECO:0000255; solute carrier family 25, member 38 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54977 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96DW6 | |
SLC25A38 | |
Synthetic peptide directed towards the middle region of human SLC25A38. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction