Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A38 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989884
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9884 Supplier Bioss Supplier No. BS19815R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A38 is a members of the solute carrier family 25 (SLC25) which is known to transport molecules over the mitochondrial membrane.

Specifications

Antigen SLC25A38
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene SLC25A38
Gene Accession No. Q96DW6
Gene Alias appoptosin; AV019094; BC010801; BOS_20700; HAMAP-Rule:MF_03064}; LOC100343972; Mitochondrial glycine transporter; Mitochondrial glycine transporter GlyC; RGD1311914; SIDBA2; Slc25a38; Solute carrier family 25 member 38; solute carrier family 25 member 38 {ECO:0000255; solute carrier family 25, member 38
Gene Symbols SLC25A38
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC25A38.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54977
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.