Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555415 Supplier Invitrogen™ Supplier No. PA555415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLQSQRPSMA SELMPSSRLW SLSYTKWKCL LYCNGVLEPL YLCPNGARCA TWFQDPTRFT GTMDAFVKIV RHEGTRTL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 80%.

SLC25A39 is a member of the solute carrier family 25 and is known to transport molecules over the mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A39
Gene Accession No. Q9BZJ4
Gene Alias 3010027G13Rik; CGI69; CGI-69; D11Ertd333e; hypothetical protein MGC63736; PRO2163; RGD1306193; Slc25a39; Solute carrier family 25 member 39; solute carrier family 25, member 39; zgc:63736
Gene Symbols SLC25A39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A39. Recombinant protein control fragment (Product #RP-89827).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51629
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.