Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542467 Supplier Invitrogen™ Supplier No. PA542467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RVNPLHVDST WLLLRRIRAE SGTKGLFAGF LPRIIKAAPS CAIMISTYEF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

SLC25A39 is a member of the solute carrier family 25 and is known to transport molecules over the mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A39
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC25A39
Gene Accession No. Q9BZJ4
Gene Alias 3010027G13Rik; CGI69; CGI-69; D11Ertd333e; hypothetical protein MGC63736; PRO2163; RGD1306193; Slc25a39; Solute carrier family 25 member 39; solute carrier family 25, member 39; zgc:63736
Gene Symbols SLC25A39
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SLC25A39 (aa 294-343).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51629
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.