Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A42 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | SLC25A42 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15960520
![]() |
Novus Biologicals
NBP15960520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159605
![]() |
Novus Biologicals
NBP159605 |
100 μL |
Each for $487.50
|
|
|||||
Description
SLC25A42 Polyclonal specifically detects SLC25A42 in Human samples. It is validated for Western Blot.Specifications
| SLC25A42 | |
| Polyclonal | |
| Rabbit | |
| Q86VD7 | |
| 284439 | |
| Synthetic peptides corresponding to SLC25A42(solute carrier family 25, member 42) The peptide sequence was selected from the N terminal of SLC25A42. Peptide sequence GNGVKEGPVRLHEDAEAVLSSSVSSKRDHRQVLSSLLPGALAGALAKTAV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| MGC26694, solute carrier family 25 member 42, solute carrier family 25, member 42 | |
| SLC25A42 | |
| IgG | |
| 35 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title