Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561975 Supplier Invitrogen™ Supplier No. PA561975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNGVKEGPVR LHEDAEAVLS SSVSSKRDHR QVLSS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 77%.

SLC25A42 belongs to the SLC25 family of mitochondrial carrier proteins.The exact function of SLC25A42 remains unknown.SLC25A42 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]. Sequence Note: removed 2 bases from the 5' end that did not align to the reference genome assembly. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-3160 BC045598.1 3-3162.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A42
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A42
Gene Accession No. Q86VD7
Gene Alias mitochondrial coenzyme A transporter SLC25A42; SLC25A42; Solute carrier family 25 member 42
Gene Symbols SLC25A42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A42. Recombinant protein control fragment (Product #RP-99214).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.