Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566772 Supplier Invitrogen™ Supplier No. PA566772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVSQHLMMQRK GEKMGRFQVR GNPEGQGVVA FGQTKDIIRQ ILQADGLRGF Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

SLC25A44 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; transmembrane transporter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A44
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A44
Gene Accession No. Q96H78
Gene Alias 6720482A19; B430110G05Rik; KIAA0446; LOW QUALITY PROTEIN: solute carrier family 25 member 44; RGD1307244; SLC25A44; Solute carrier family 25 member 44; solute carrier family 25, member 44
Gene Symbols SLC25A44
Host Species Rabbit
Immunogen Recombinant Human SLC25A44. Recombinant protein control fragment (Product #RP-107429).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.