Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A45 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562251
Description
Immunogen sequence: FDLIKVRLQN QTEPRAQPGS PPPRYQGPVH CAASIFREEG PRGLFRGAWA LTLRDTPTVG IYFITYEGLC RQYTPEGQN Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
The function remains unknown.
Specifications
SLC25A45 | |
Polyclonal | |
Unconjugated | |
SLC25A45 | |
AW491445; LOW QUALITY PROTEIN: solute carrier family 25 member 45; SLC25A45; Solute carrier family 25 member 45; solute carrier family 25, member 45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283130 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N413 | |
SLC25A45 | |
Recombinant protein corresponding to Human SLC25A45. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction