Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598791
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598791 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598791 Supplier Invitrogen™ Supplier No. PA598791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A46 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

SLC25A46 belongs to the SLC25 family of mitochondrial carrier proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A46
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC25A46
Gene Accession No. Q96AG3, Q9CQS4
Gene Alias 1200007B05Rik; AI325987; HMSN6B; RGD1305072; Slc25a46; Solute carrier family 25 member 46; solute carrier family 25, member 46; TB1
Gene Symbols SLC25A46
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 25 member 46 protein (1-102AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67453, 91137
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.