Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A53 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569333
Description
This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.
SLC25A53 (Solute Carrier Family 25 Member 53) is a Protein Coding gene.
Specifications
SLC25A53 | |
Polyclonal | |
Unconjugated | |
SLC25A53 | |
2310046F18Rik; 2810402A17Rik; AI448995; AI841128; Mcart1l; MCART6; mitochondrial carrier triple repeat 1-like; mitochondrial carrier triple repeat 6; mitochondrial carrier triple repeat protein 6; Slc25a53; Solute carrier family 25 member 53; solute carrier family 25, member 53 | |
Rabbit | |
Affinity chromatography | |
RUO | |
401612 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5H9E4 | |
SLC25A53 | |
synthetic peptide directed towards the middle region of human SLC25A53. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction