Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A53 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571718
Description
The function of this protein remains unknown.
Specifications
SLC25A53 | |
Polyclonal | |
Unconjugated | |
SLC25A53 | |
Solute carrier family 25 member 53; Mitochondrial carrier triple repeat protein 6; SLC25A53; MCART6 | |
Rabbit | |
Protein A | |
RUO | |
401612 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q5H9E4 | |
SLC25A53 | |
KLH conjugated synthetic peptide between 279-307 amino acids from the C-terminal region of human MCART6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction