Learn More
Invitrogen™ SLC25A6 Polyclonal Antibody

Description
Recommended positive controls: 293T. Predicted reactivity: Zebrafish (93%), Pig (96%), Chicken (93%), Sheep (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SLC25A6 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Slc25a6 |
| Gene Accession No. | P12236, Q5U680 |
| Gene Alias | 4930433D19Rik; 4933433F13Rik; AAC3; adenine nucleotide translocator 2; adenine nucleotide translocator 3; adenine nucleotide translocator 3 (liver); ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocase 3, N-terminally processed; ADP/ATP translocator of liver; ANT; ANT 2; ANT 3; ANT3; ANT3Y; AW557176; CDABP0051; D6Bwg0781e; mitochondrial ADP/ATP carrier protein; mitochondrial S-adenosylmethionine transporter; mitochondrial solute carrier family 25 member 6; S-adenosylmethionine mitochondrial carrier protein; Samc; si:dkey-21o13.4; Slc25a26; slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; solute carrier family 25 member 26; Solute carrier family 25 member 6; solute carrier family 25, member A6; translocase; wu:fj78b08 |
| Gene Symbols | Slc25a6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.