Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529967
Description
Recommended positive controls: Jurkat, Raji, NCI-H929. Predicted reactivity: Zebrafish (91%), Pig (95%), Chicken (93%), Sheep (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.
Specifications
SLC25A6 | |
Polyclonal | |
Unconjugated | |
Slc25a6 | |
4930433D19Rik; 4933433F13Rik; AAC3; adenine nucleotide translocator 2; adenine nucleotide translocator 3; adenine nucleotide translocator 3 (liver); ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocase 3, N-terminally processed; ADP/ATP translocator of liver; ANT; ANT 2; ANT 3; ANT3; ANT3Y; AW557176; CDABP0051; D6Bwg0781e; mitochondrial ADP/ATP carrier protein; mitochondrial S-adenosylmethionine transporter; mitochondrial solute carrier family 25 member 6; S-adenosylmethionine mitochondrial carrier protein; Samc; si:dkey-21o13.4; Slc25a26; slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; solute carrier family 25 member 26; Solute carrier family 25 member 6; solute carrier family 25, member A6; translocase; wu:fj78b08 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P12236 | |
Slc25a6 | |
Recombinant fragment corresponding to a region within amino acids 57 and 298 of Human SLC25A6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction