Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575458 Supplier Invitrogen™ Supplier No. PA575458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc25a6
Gene Accession No. P12236, Q5U680
Gene Alias 4930433D19Rik; 4933433F13Rik; AAC3; adenine nucleotide translocator 2; adenine nucleotide translocator 3; adenine nucleotide translocator 3 (liver); ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocase 3, N-terminally processed; ADP/ATP translocator of liver; ANT; ANT 2; ANT 3; ANT3; ANT3Y; AW557176; CDABP0051; D6Bwg0781e; mitochondrial ADP/ATP carrier protein; mitochondrial S-adenosylmethionine transporter; mitochondrial solute carrier family 25 member 6; S-adenosylmethionine mitochondrial carrier protein; Samc; si:dkey-21o13.4; Slc25a26; slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; solute carrier family 25 member 26; Solute carrier family 25 member 6; solute carrier family 25, member A6; translocase; wu:fj78b08
Gene Symbols Slc25a6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 120-160 of Human SLC25A6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293, 498885, 67582
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.