Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542488 Supplier Invitrogen™ Supplier No. PA542488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ACCSPPVRDI LSRGGFLGEG PGDTAEEEQL FLSVHDAVQT ARARHRELEA Sequence homology: Cow: 79%; Dog: 79%; Horse: 79%; Human: 100%.

SLC26A1 is a member of sulfate/anion transporter family. Family members are well conserved in their protein (aa length among species) structures, but have markedly different tissue expression patterns. Its gene is primarily expressed in the liver, pancreas, and brain. This gene is a member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures, but have markedly different tissue expression patterns. This gene is primarily expressed in the liver, pancreas, and brain. Three splice variants that encode different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC26A1
Gene Accession No. Q9H2B4
Gene Alias CAON; EDM4; SAT1; SAT-1; SLC26A1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; solute carrier family 26 member 1; solute carrier family 26, member 1; sulfate anion tranporter AT1; sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein
Gene Symbols SLC26A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC26A1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10861
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.