Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116742 Supplier Invitrogen™ Supplier No. PA5116742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: mouse heart, mouse brain Immunogen sequence: EILSRALEVS PPRCLVLECT HVCSIDYTVV LGLGELLQDF QKQGVALAFV GLQVPVLRVL LSADLKGFQY FSTLEEAEKH LRQEPGTQPY NIREDSILDQ K.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC26A11
Gene Accession No. Q80ZD3
Gene Alias anion exchanger Slc26a11; F630021I08Rik; novel sulfate transporter protein (Slc26a11); slc26a11; Sodium-independent sulfate anion transporter; solute carrier family 26 (anion exchanger), member 11; solute carrier family 26 member 11; solute carrier family 26, member 11
Gene Symbols SLC26A11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500-600 of human SLC26A11 (NP_0011598191).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268512
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.