Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563524 Supplier Invitrogen™ Supplier No. PA563524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESKEQHNVSP RDSAEGNDSY PSGIHLELQR ESSTDFKQFE TNDQCRPYHR ILIERQEKSD TNFKEFVIKK LQKNCQCSP Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 51%.

The SLC26 family is comprised of sulfate/anion transporters that are well conserved in both their genomic and protein structures, yet have markedly different tissue expression patterns. Members of the SLC26 family can mediate the electroneutral exchange of Cl- for HCO3- across the plasma membrane of mammalian cells. SLC26A2 (solute carrier family 26, member 2), also known as DTD, EDM4, DTDST, MST153, D5S1708 or MSTP157, is a 739 amino acid diastrophic dysplasia sulfate transporter belonging to the SLC26A/SulP transporter family. Ubiquitously expressed, SLC26A2 may play a role in endochondral bone formation. SLC26A2 is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias and is critical for sulfation of proteoglycans and matrix organization in cartilage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC26A2
Gene Accession No. P50443
Gene Alias D5S1708; Diastrophic dysplasia protein; diastrophic dysplasia protein homolog; Dtd; DTD sulfate transporter; DTDST; EDM4; MST153; MSTP157; SLC26A2; solute carrier family 26 (anion exchanger), member 2; solute carrier family 26 (sulfate transporter), member 2; solute carrier family 26 member 2; solute carrier protein 26a2; ST-OB; sulfate anion transporter 1; sulfate transporter
Gene Symbols SLC26A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC26A2. Recombinant protein control fragment (Product #RP-103884).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.