Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC26A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110385
Description
Immunogen sequence: MSSESKEQHN VSPRDSAEGN DSYPSGIHLE LQRESSTDFK QFETNDQCRP YHRILIERQE KSDTNFKEFV IKKLQKNCQC SPAKAKNMIL GFLPVLQWLP KYDLKKNILG D.
The SLC26 family is comprised of sulfate/anion transporters that are well conserved in both their genomic and protein structures, yet have markedly different tissue expression patterns. Members of the SLC26 family can mediate the electroneutral exchange of Cl- for HCO3- across the plasma membrane of mammalian cells. SLC26A2 (solute carrier family 26, member 2), also known as DTD, EDM4, DTDST, MST153, D5S1708 or MSTP157, is a 739 amino acid diastrophic dysplasia sulfate transporter belonging to the SLC26A/SulP transporter family. Ubiquitously expressed, SLC26A2 may play a role in endochondral bone formation. SLC26A2 is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias and is critical for sulfation of proteoglycans and matrix organization in cartilage.
Specifications
SLC26A2 | |
Polyclonal | |
Unconjugated | |
SLC26A2 | |
D5S1708; Diastrophic dysplasia protein; diastrophic dysplasia protein homolog; Dtd; DTD sulfate transporter; DTDST; EDM4; MST153; MSTP157; SLC26A2; solute carrier family 26 (anion exchanger), member 2; solute carrier family 26 (sulfate transporter), member 2; solute carrier family 26 member 2; solute carrier protein 26a2; ST-OB; sulfate anion transporter 1; sulfate transporter | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117267, 13521, 1836 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O70531, P50443, Q62273 | |
SLC26A2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human SLC26A2 (NP_000103.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction