Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC26A2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7796 Supplier Bethyl Laboratories, Inc Supplier No. A304467A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The SLC26 family is comprised of sulfate/anion transporters that are well conserved in both their genomic and protein structures, yet have markedly different tissue expression patterns. Members of the SLC26 family can mediate the electroneutral exchange of Cl- for HCO3- across the plasma membrane of mammalian cells. SLC26A2 (solute carrier family 26, member 2), also known as DTD, EDM4, DTDST, MST153, D5S1708 or MSTP157, is a 739 amino acid diastrophic dysplasia sulfate transporter belonging to the SLC26A/SulP transporter family. Ubiquitously expressed, SLC26A2 may play a role in endochondral bone formation. SLC26A2 is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias and is critical for sulfation of proteoglycans and matrix organization in cartilage.

Specifications

Antigen SLC26A2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SLC26A2
Gene Accession No. P50443
Gene Alias D5S1708, diastrophic dysplasia protein, DTD, DTDST, EDM4, MST153, MSTP157, solute carrier family 26 (anion exchanger), member 2, solute carrier family 26 (sulfate transporter), member 2, Solute carrier family 26 member 2, sulfate anion transporter 1
Gene Symbols SLC26A2
Host Species Rabbit
Immunogen Between 689 and 739
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1836
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.