Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568530 Supplier Invitrogen™ Supplier No. PA568530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 79%; Guinea Pig: 79%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 86%; Rat: 93%; Sheep: 100%.

SLC26A3 is a chloride/bicarbonate exchanger that is involved in absorption inside the colon. SLC26A3 helps mediate electrolyte and fluid absorption. The protein encoded by the SLC26A3 gene is a transmembrane glycoprotein that functions as a sulfate transporter, and it is localized to the mucosa. The chloride ions transported by SLC26A3 cross the cell membrane in exchange for biocarbonate ions. SLC26A3 is localized in the mucosa of the lower intestinal tract, specifically the apical membrane of the columnar epithelium and in some goblet cells. Diseases associated with SLC26A3 protein dysfunction include congenital chloride diarrhea and congenital secretory chloride diarrhea 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC26A3
Gene Accession No. P40879
Gene Alias 9030623B18Rik; 9130013M11Rik; anion exchange protein; AV376035; chloride anion exchanger; CLD; down-regulated in adenoma; down-regulated in adenoma DRA; down-regulated in adenoma protein; DRA; Protein DRA; Slc26a3; solute carrier family 26 (anion exchanger), member 3; solute carrier family 26 member 3; solute carrier family 26, member 3
Gene Symbols SLC26A3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC26A3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1811
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.