Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598158
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598158 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598158 Supplier Invitrogen™ Supplier No. PA598158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC26A4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Mutations in this gene are associated with Pendred syndrome, the most common form of syndromic deafness, an autosomal-recessive disease. It is highly homologous to the SLC26A3 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC26A4
Gene Accession No. O43511
Gene Alias DFNB4; EVA; PDS; Pendred syndrome homolog; Pendred's syndrome; pendrin; Slc26a4; Sodium-independent chloride/iodide transporter; solute carrier family 26 (anion exchanger), member 4; solute carrier family 26 member 4; solute carrier family 26, member 4; TDH2B; truncated solute carrier family 26
Gene Symbols SLC26A4
Host Species Rabbit
Immunogen Recombinant Human Pendrin protein (592-655AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5172
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.