Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC26A4 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30394020UL
Description
SLC26A4 Polyclonal specifically detects SLC26A4 in Mouse samples. It is validated for Western Blot.Specifications
SLC26A4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
5172 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
DFNB4, EVA, PDSTDH2B, pendrin, Sodium-independent chloride/iodide transporter, Solute carrier family 26 member 4, solute carrier family 26, member 4 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human SLC26A4 (NP_000432.1). | |
20 μL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction