Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC26A5 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP128795

Catalog No. NBP128795


Only null left
Add to Cart

Description

Description

SLC26A5 Polyclonal specifically detects SLC26A5 in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

SLC26A5
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
DFNB61PRESprestin (motor protein), MGC118886, MGC118887, MGC118888, MGC118889, prestin, Solute carrier family 26 member 5, solute carrier family 26, member 5 (prestin)
Goat
Affinity Purified
RUO
375611
Human
IgG
ELISA
0.5 mg/mL
Peptide ELISA Detection limit 1:32000
NP_945350.1
SLC26A5
Peptide with sequence ERLHTKDKVPDSIAD -C corresponding to N-Terminus (near) according to NP_945350.1, NP_996766.1, NP_996767.1, NP_996768.1.
0.1 mg
Primary
This antibody is expected to recognize reported isoforms NP_996768.1; NP_996767.1; NP_996766.1; NP_945350.1.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only