Learn More
Invitrogen™ SLC26A6 Polyclonal Antibody

Description
Immunogen sequence: ATVYFANAEF YSDALKQRCG VDVDFLISQK KKLLKKQEQL KLKQLQKEEK LRKQAASPKG ASVSINVNTS LEDMRSNNVE DCKMMQVS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 59%.
Specifications
Specifications
| Antigen | SLC26A6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC26A6 |
| Gene Accession No. | Q9BXS9 |
| Gene Alias | anion exchange transporter; anion transporter 1; B930010B04Rik; CFEX; Chloride-formate exchanger; DKFZp586E1422; Pat1; Pat-1; pendrin L1; pendrin-L1; Pendrin-like protein 1; putative anion transporter-1; SLC26A6; solute carrier family 26 (anion exchanger), member 6; solute carrier family 26 member 6; solute carrier family 26, member 6; sulfate anion transporter |
| Gene Symbols | SLC26A6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.