Learn More
Invitrogen™ SLC26A6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110386
Description
Immunogen sequence: QMPHYSVLGQ VPDTDIYRDV AEYSEAKEVR GVKVFRSSAT VYFANAEFYS DALKQRCGVD VDFLISQKKK LLKKQEQLKL KQLQKEEKLR KQAASPKGAS VSINVNTSLE DMRSNNVEDC KMMVSSGDKM EDATANGQED SKAPDGSTLK ALGLPQPDFH SLILDLGALS FVDTVCLKSL KNIFHDFREI EVEVYMAACH SPVVSQLEAG HFFDASITKK HLFASVHDAV TFALQHPRPV PDSPVSVTRL.
This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
SLC26A6 | |
Polyclonal | |
Unconjugated | |
SLC26A6 | |
anion exchange transporter; anion transporter 1; B930010B04Rik; CFEX; Chloride-formate exchanger; DKFZp586E1422; Pat1; Pat-1; pendrin L1; pendrin-L1; Pendrin-like protein 1; putative anion transporter-1; SLC26A6; solute carrier family 26 (anion exchanger), member 6; solute carrier family 26 member 6; solute carrier family 26, member 6; sulfate anion transporter | |
Rabbit | |
Affinity Chromatography | |
RUO | |
65010 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BXS9 | |
SLC26A6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 509-758 of human SLC26A6 (NP_599025.2). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.