Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC26A6 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537970
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| SLC26A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC26A6 | |
| anion exchange transporter; anion transporter 1; B930010B04Rik; CFEX; Chloride-formate exchanger; DKFZp586E1422; Pat1; Pat-1; pendrin L1; pendrin-L1; Pendrin-like protein 1; putative anion transporter-1; SLC26A6; solute carrier family 26 (anion exchanger), member 6; solute carrier family 26 member 6; solute carrier family 26, member 6; sulfate anion transporter | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 65010 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BXS9 | |
| SLC26A6 | |
| Peptide with sequence C-SAPRTHQWRT, from the internal region of SLC26A6 (aa 49-59). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction