Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC26A8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119457
This item is not returnable.
View return policy
Description
SLC26A8 Polyclonal Antibody for Western Blot, IHC (P)
SLC26A8 is one member of a family of sulfate/anion transporters. Family members are well conserved in their protein (aa length among species) structures yet have markedly different tissue expression patterns.
Specifications
SLC26A8 | |
Polyclonal | |
Unconjugated | |
SLC26A8 | |
Anion exchange transporter; anion transporter/exchanger-8; RP11-482O9.1; SLC26A8; solute carrier family 26 (anion exchanger), member 8; solute carrier family 26 member 8; SPGF3; TAT1; testis anion transporter 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
116369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q96RN1 | |
SLC26A8 | |
Recombinant protein Testis Anion Transporter 1. The antigen corresponds to amino acid range 664-970 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction