Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC26A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121604
This item is not returnable.
View return policy
Description
SLC26A8 Polyclonal Antibody for Western Blot, IHC (P)
SLC26A8 is one member of a family of sulfate/anion transporters. Family members are well conserved in their protein (aa length among species) structures yet have markedly different tissue expression patterns.
Specifications
SLC26A8 | |
Polyclonal | |
Unconjugated | |
SLC26A8 | |
Anion exchange transporter; anion transporter/exchanger-8; RP11-482O9.1; SLC26A8; solute carrier family 26 (anion exchanger), member 8; solute carrier family 26 member 8; SPGF3; TAT1; testis anion transporter 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
116369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q96RN1 | |
SLC26A8 | |
Recombinant protein Testis Anion Transporter 1. The antigen corresponds to amino acid range 664-970 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction