Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558240 Supplier Invitrogen™ Supplier No. PA558240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELEPEMEPKA ETETKTQTEM EPQPETEPEM EPNPKSRPRA HTFPQQRYWP MYHPSMASTQ SQTQTRTWSV ERRRHPMDSY SPEGNSNEDV Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 30%.

SLC26A8 is one member of a family of sulfate/anion transporters. Family members are well conserved in their protein (aa length among species) structures yet have markedly different tissue expression patterns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC26A8
Gene Accession No. Q96RN1
Gene Alias Anion exchange transporter; anion transporter/exchanger-8; RP11-482O9.1; SLC26A8; solute carrier family 26 (anion exchanger), member 8; solute carrier family 26 member 8; SPGF3; TAT1; testis anion transporter 1
Gene Symbols SLC26A8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC26A8. Recombinant protein control fragment (Product #RP-95608).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116369
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.