Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC26A9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542506
Description
Peptide sequence: LAKLSSTYGK IGVKVFLVNI HAQVYNDISH GGVFEDGSLE CKHVFPSIHD Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 85%.
This gene is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns.
Specifications
SLC26A9 | |
Polyclonal | |
Unconjugated | |
SLC26A9 | |
Anion transporter/exchanger protein 9; anion transporter/exchanger-9; E030002L01Rik; SLC26A9; SLC26A9 anion transporter/exchanger; solute carrier family 26 (anion exchanger), member 9; solute carrier family 26 member 9; solute carrier family 26, member 9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
115019 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7LBE3 | |
SLC26A9 | |
Synthetic peptide directed towards the middle region of human SLC26A9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction