Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC27A1 Polyclonal Antibody

SLC27A1 Polyclonal Antibody

Supplier:  Thermo Scientific PA550574

Catalog No. PA550574


Only null left
Add to Cart

Description

Description

SLC27A1 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC27A1
Polyclonal
Unconjugated
SLC27A1
ACSVL5; ACSVL5FATP-1; EC 6.2.1; Fatp; FATP1; FATP-1; Fatty acid transport protein; fatty acid transport protein 1; FLJ00336; long-chain fatty acid transport protein 1; MGC71751; SLC27A1; solute carrier family 27 (fatty acid transporter), member 1; solute carrier family 27 member 1; solute carrier family 27, member 1
Rabbit
Antigen Affinity Chromatography
RUO
376497
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6PCB7
SLC27A1
Synthetic peptide corresponding to residues near the C terminal of human solute carrier family 27 (fatty acid transporter), member 1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.