Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC27A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585002 Supplier Invitrogen™ Supplier No. PA585002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QREGFDPRQT SDRLFFLDLK QGHYLPLNEA VYTRICSGAF Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 80%.

SLC27A1 is a protein coding gene. Mediates the ATP-dependent import of long-chain fatty acids (LCFA) into the cell by mediating their translocation at the plasma membrane. Has also an acyl-CoA ligase activity for long-chain and very-long-chain fatty acids. May act directly as a bona fide transporter, or alternatively, in a cytoplasmic or membrane-associated multimeric protein complex to trap and draw fatty acids towards accumulation. Plays a pivotal role in regulating available LCFA substrates from exogenous sources in tissues undergoing high levels of beta-oxidation or triglyceride synthesis. May be involved in regulation of cholesterol metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC27A1
Gene Accession No. Q6PCB7
Gene Alias ACSVL5; ACSVL5FATP-1; EC 6.2.1; Fatp; FATP1; FATP-1; Fatty acid transport protein; fatty acid transport protein 1; FLJ00336; long-chain fatty acid transport protein 1; MGC71751; SLC27A1; solute carrier family 27 (fatty acid transporter), member 1; solute carrier family 27 member 1; solute carrier family 27, member 1
Gene Symbols SLC27A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC27A1. Recombinant protein control fragment (Product #RP-108661).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 376497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.