Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC27A2 Mouse anti-Human, Clone:6B3A9, Abnova™
Mouse monoclonal antibody raised against recombinant human SLC27A2.
Supplier: Abnova Corporation MAB17766
Description
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes, but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]Specifications
SLC27A2 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry Immunocytochemistry Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
SLC27A2 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
6B3A9 | |
solute carrier family 27 (fatty acid transporter), member 2 | |
In PBS (0.05% sodium azide) | |
ACSVL1, FACVL1, FATP2, HsT17226, VLACS, VLCS, hFACVL1 | |
Recombinant protein corresponding to amino acids 346-405 of human SLC27A2 from E. coli. | |
11001 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction