Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC27A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102343
Description
Antibody detects endogenous levels of total SLC27A2.
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy.
Specifications
SLC27A2 | |
Polyclonal | |
Unconjugated | |
SLC27A2 | |
ACSVL1; Arachidonate--CoA ligase; Facvl1; FATP2; FATP-2; Fatty acid transport protein 2; fatty acid transporter; fatty-acid-coenzyme A ligase, very long-chain 1; hFACVL1; HsT17226; Long-chain-fatty-acid--CoA ligase; Phytanate--CoA ligase; SLC27A2; solute carrier family 27 (fatty acid transporter) member 2; solute carrier family 27 (fatty acid transporter), member 2; solute carrier family 27 (fatty acid transporter), member 32; solute carrier family 27 member 2; THCA-CoA ligase; ve; very long-chain acyl-CoA synthetase; very long-chain acyl-Coenzyme A dehydrogenase synthase; very long-chain fatty-acid-coenzyme A ligase 1; very long-chain-fatty-acid-CoA ligase; very-long-chain acyl-CoA synthetase; Vlac; VLACS; Vlcs | |
Rabbit | |
Affinity chromatography | |
RUO | |
11001, 26458, 65192 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O14975, O35488, P97524 | |
SLC27A2 | |
A synthesized peptide derived from human SLC27A2(Accession O14975), corresponding to amino acid residues E61-M111. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction