Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC27A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542429 Supplier Invitrogen™ Supplier No. PA542429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LLFRDETLTY AQVDRRSNQV ARALHDHLGL RQGDCVALLM GNEPAYVWLW Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC27A2
Gene Accession No. O14975
Gene Alias ACSVL1; Arachidonate--CoA ligase; Facvl1; FATP2; FATP-2; Fatty acid transport protein 2; fatty acid transporter; fatty-acid-coenzyme A ligase, very long-chain 1; hFACVL1; HsT17226; Long-chain-fatty-acid--CoA ligase; Phytanate--CoA ligase; SLC27A2; solute carrier family 27 (fatty acid transporter) member 2; solute carrier family 27 (fatty acid transporter), member 2; solute carrier family 27 (fatty acid transporter), member 32; solute carrier family 27 member 2; THCA-CoA ligase; ve; very long-chain acyl-CoA synthetase; very long-chain acyl-Coenzyme A dehydrogenase synthase; very long-chain fatty-acid-coenzyme A ligase 1; very long-chain-fatty-acid-CoA ligase; very-long-chain acyl-CoA synthetase; Vlac; VLACS; Vlcs
Gene Symbols SLC27A2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC27A2 (aa 72-121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11001
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.