Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC27A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552364
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552364 Supplier Invitrogen™ Supplier No. PA552364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEAERESNRA ARAFLRALGW DWGPDGGDSG EGSAGEGERA APGAGDAAAG SGAEFAGGDG AARGGGAAAP LSPGATVALL LPAGPEFLWL WFGLAKAGLR TAFVPTALRR GPLLHCLRSC GARALVLAPG KAGAP Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.

SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC27A3
Gene Accession No. Q5K4L6
Gene Alias Acsvl3; FATP3; FATP-3; fatty acid transport protein 3; long-chain fatty acid transport protein 3; mFLJ00207 protein; PSEC0067; Slc27a3; solute carrier family 27 (fatty acid transporter), member 3; solute carrier family 27 member 3; UNQ367/PRO703; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3
Gene Symbols SLC27A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC27A3. Recombinant protein control fragment (Product #RP-89173).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.