Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC27A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566931
Description
Immunogen sequence: DTWERFVRRFG PLQVLETYGL TEGNVATINY TGQRGAVGRA SWLYKHIFPF SLIRYDVTTG EPIRDPQGHC MATSPGEPGL LVAPVSQQSP FLGYAGGPEL AQGKLLKDVF R Highest antigen sequence identity to the following orthologs - mouse 88%, rat 89%.
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
Specifications
SLC27A3 | |
Polyclonal | |
Unconjugated | |
SLC27A3 | |
Acsvl3; FATP3; FATP-3; fatty acid transport protein 3; long-chain fatty acid transport protein 3; mFLJ00207 protein; PSEC0067; Slc27a3; solute carrier family 27 (fatty acid transporter), member 3; solute carrier family 27 member 3; UNQ367/PRO703; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11000 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5K4L6 | |
SLC27A3 | |
Recombinant Human SLC27A3. Recombinant protein control fragment (Product #RP-107584). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction